Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD30 Mouse anti-Human, Clone: Ber-H2, Cocktail, Zeta

Catalog No. 502215895
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
7 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-221-5895 7 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-221-5895 Supplier ZETA CORPORATION Supplier No. Z2101MP
Add to Cart
Add to Cart

Mouse Monoclonal Antibody Cocktail

This product is diluted and in a ready-to-use formulation. A recommended positive control tissue for this product is Hodgkin lymphoma, however positive controls are not limited to this tissue type. The primary antibody is intended for laboratory professional use in the detection of the corresponding protein in formalin-fixed, paraffin-embedded tissue stained in manual qualitative immunohistochemistry (IHC) testing. This antibody is intended to be used after the primary diagnosis of tumor has been made by conventional histopathology using non-immunological histochemical stains.

CD30 Monoclonal antibody specifically detects CD30 in Human samples. It is validated for Immunohistochemistry (Paraffin)

Specifications

Antigen CD30
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone Ber-H2
Conjugate Unconjugated
Formulation tris with BSA, NP-40 and <0.1% sodium azide
Gene TNFRSF8
Gene Accession No. P28908
Gene Alias Ber-H2; Cd30; CD30 antigen; CD30 molecule; CD30L receptor; cytokine receptor CD30; D1S166E; Ki; Ki-1; Ki-1 antigen; Lymphocyte activation antigen CD30; sCD30; soluble CD30; TNF receptor superfamily member 8; TNFRSF8; Tumor necrosis factor receptor superfamily member 8; tumor necrosis factor receptor superfamily, member 8; Tumor necrosis factor superfamily member 8; tumor necrosis factor superfamily, member CD30
Host Species Mouse
Immunogen Co cell line established from a patient with Hodgkin's disease of T-cell lineage
Purification Method Protein A
Quantity 7 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 943
Target Species Human
Content And Storage 2-8°C
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.