Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD30 Mouse anti-Human, FITC, Clone: Ber-H83, BD™
Mouse Monoclonal Antibody
Supplier: BD Biosciences 341644
Description
CD30 Monoclonal antibody specifically detects CD30 in Human samples. It is validated for Flow cytometry.
Specifications
CD30 | |
Monoclonal | |
3 µg/mL | |
Vol. Per test: 20μL | |
Mouse | |
50 Tests | |
Cell Biology | |
943 | |
IgG1 κ |
Flow Cytometry | |
Ber-H83 | |
FITC | |
Phosphate buffered saline with gelatin and 0.1% sodium azide. | |
Co Cells | |
RUO | |
Primary | |
Human | |
Purified |
Safety and Handling
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Reseach Use Only.
Spot an opportunity for improvement?Share a Content Correction