Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD30 Mouse anti-Human, Unconjugated, Clone: CON6D/B5, MilliporeSigma™

Catalog No. MABF19931MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABF19931MI 100 μg
MABF19932MI 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MABF19931MI Supplier MilliporeSigma Supplier No. MABF1993100UG
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

CD30 Monoclonal antibody specifically detects CD30 in Human samples. It is validated for Immunohistochemistry (Paraffin).
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD30
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone CON6D/B5
Conjugate Unconjugated
Gene Accession No. NP_001234
Gene Alias Tumor necrosis factor receptor superfamily member;CD30L receptor;Ki-1 antigen;Lymphocyte activation antigen CD30
Gene Symbols TNFRSF8, CD30
Host Species Mouse
Immunogen L428 Hodgkin's lymphoma cells.
Molecular Weight of Antigen 63.75 kDa calculated.
Purification Method Protein G Purified
Quantity 100 μg
Research Discipline Inflammation and Immunology
Primary or Secondary Primary
Test Specificity Clone CON6D/B5 is a mouse monoclonal antibody that detects human CD30. It targets an epitope within the extracellular domain.
Target Species Human
Content And Storage Stable for 1 year at 2 to 8°C from date of receipt.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.