Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD30 Rabbit anti-Human, Polyclonal, Proteintech
SDP

Catalog No. 501736006 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-173-6006 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-173-6006 Supplier Proteintech Group Inc Supplier No. 551321AP150UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

GTP exchange factor, RCC1, with chromatin , but Ran is shown here to bind directly to chromatin in two modes, either dependent or independent of RCC1, and, where bound, to increase the affinity of chromatin for NE membranes.

Specifications

Antigen CD30
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.13 mg/mL
Conjugate Unconjugated
Formulation 0.1 M glycine, 0.1 M sodium bicarbonate with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene TNFRSF8
Gene Accession No. P28908
Gene Alias CD30, CD30L receptor, D1S166E, Ki 1, Ki 1 antigen, TNFRSF8
Gene Symbols TNFRSF8
Host Species Rabbit
Immunogen Peptide
Purification Method Antigen Affinity Chromatography
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 943
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.