Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Epredia™ Lab Vision™ CD30 (Reed-Sternberg Cell Marker) Ab-1, Mouse Monoclonal Antibody

Catalog No. MS361S0 Shop All Epredia Products
Change view
Click to view available options
Quantity:
100 μL
500 μL
1 mL
7 mL
12 mL
Form:
Concentrated
Prediluted
Ready-to-Use
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Form
MS361S0 100 μL Concentrated
MS361S 1 mL Concentrated
MS361R7 7 mL Prediluted
MS361RQ 12 mL Ready-to-Use
MS361S1 500 μL Concentrated
MS361R1 1 mL Prediluted
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. MS361S0 Supplier Epredia™ Supplier No. MS361S0
Add to Cart
Add to Cart

Obtain accurate and reproducible results in immunohistochemistry experiments with Epredia™ CD30 (Reed-Sternberg Cell Marker) Ab-1, Mouse Monoclonal Antibody.

CD30, a single chain glycoprotein, is synthesized as a 90 kDa precursor which is processed in the Golgi complex into a membrane-bound phosphorylated mature 105/120kDa glycoprotein. The CD30/Ki-1 antigen is expressed by mononuclear Hodgkin and multinucleated Reed-Sternberg cells in Hodgkin's disease, by the tumor cells of a majority of anaplastic large cell lymphomas, and by a varying proportion of activated T and B cells.

Host Species: Mouse

Clone: Ber-H2

Isotype: IgG1/κ

Species Reactivity: Human. Does not react with baboon and monkey. Others not known.

Epitope: Not determined

Immunogen: Co cells

Molecular Weight: 120 kDa (mature), 95 kDa (precursor)

Positive Control: Hodgkin's lymphoma

Cellular Localization: Membrane and cytoplasm

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)

Specifications

Antigen CD30 Ab-1
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone Ber-H2
Conjugate Unconjugated
Host Species Mouse
Immunogen Co cells
Quantity 100 μL
Regulatory Status IVD
Research Discipline Oncogenesis
Primary or Secondary Primary
Target Species Human
Form Concentrated
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.