Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD30 (Reed-Sternberg Cell Marker) Ab-4 Mouse Monoclonal Antibody, Epredia™

Catalog No. MS1857R7 Shop All Epredia Products
Change view
Click to view available options
Quantity:
100 μL
500 μL
1 mL
7 mL
Form:
Concentrated
Prediluted
Concentration:
200μg/mL
Prediluted
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Form Concentration
MS1857R7 7 mL Prediluted Prediluted
MS1857P 1 mL Concentrated 200μg/mL
MS1857P0 100 μL Concentrated 200μg/mL
MS1857P1 500 μL Concentrated 200μg/mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. MS1857R7 Supplier Epredia Supplier No. MS1857R7

Ensure accurate, reproducible results in immunohistochemistry experiments with Epredia™ CD30 (Reed-Sternberg Cell Marker) Ab-4, Mouse Monoclonal Antibody.

CD30, a single chain glycoprotein, is synthesized as a 90 kDa precursor which is processed in the Golgi complex into a membrane-bound phosphorylated mature 105/120kDa glycoprotein. The CD30/Ki-1 antigen is expressed by mononuclear Hodgkin and multinucleated Reed-Sternberg cells in Hodgkin's disease, by the tumor cells of a majority of anaplastic large cell lymphomas, and by a varying proportion of activated T and B cells. About one third of the Ki-1 positive lymphomas lack the leukocyte common antigen (CD45).

Host Species: Mouse

Clone: HRS4

Isotype: IgG1

Species Reactivity: Human. Others not known.

Epitope: Extracellular domain

Immunogen: Recombinant extracellular domain of CD30 protein

Molecular Weight: 120 kDa (mature), 95 kDa (precursor)

Positive Control: Hodgkin's lymphoma

Cellular Localization: Membrane

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)

Specifications

Antigen CD30 Ab-4
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone HRS4
Concentration Prediluted
Conjugate Unconjugated
Host Species Mouse
Immunogen Recombinant extracellular domain of CD30 protein
Quantity 7 mL
Regulatory Status IVD
Research Discipline Oncogenesis
Primary or Secondary Primary
Target Species Human
Form Prediluted
Isotype IgG1
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.