Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD300a/LMIR1 Antibody (172224) [Biotin], Novus Biologicals™

Rat Monoclonal Antibody
Supplier: Novus Biologicals FAB1186B
Description
CD300a/LMIR1 Monoclonal antibody specifically detects CD300a/LMIR1 in Mouse samples. It is validated for Flow Cytometry, CyTOFSpecifications
CD300a/LMIR1 | |
Monoclonal | |
Biotin | |
PBS | |
Rat | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
Primary | |
Mouse | |
Purified |
Flow Cytometry, CyTOF | |
172224 | |
Flow Cytometry, CyTOF-ready | |
CD300a antigenCMRF35H9, CD300a molecule, CLM-8, CMRF-35H, CMRF35-H, CMRF35H leukocyte immunoglobulin-like receptor, CMRF35-H9, CMRF-35-H9CD300 antigen-like family member A, CMRF35HIRp60, CMRF35-like molecule 8, IgSF12, IGSF12NK inhibitory receptor, Immunoglobulin superfamily member 12, Inhibitory receptor protein 60, IRC1, IRC1/IRC2, IRC2, Irp60, leukocyte membrane antigen | |
Mouse myeloma cell line NS0-derived recombinant mouse CD300a/LMIR1, Leu28-Arg183, Accession # BAC80268 | |
0.1 mL | |
Mast Cell Markers | |
11314 | |
Store at 4°C in the dark. | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction