Learn More
CD300a/LMIR1 Antibody (MEM-260), Alexa Fluor™ 750, Novus Biologicals™
SDP
Mouse Monoclonal Antibody
Supplier:  Novus Biologicals NB11058726AF750
Catalog No. N1108726A70
Only null left
Add to Cart
Description
Description
CD300a/LMIR1 Monoclonal specifically detects CD300a/LMIR1 in Human samples. It is validated for Western Blot, Flow Cytometry.
Specifications
Specifications
CD300a/LMIR1
Monoclonal
Alexa Fluor 750
50 mM sodium borate with 0.05% sodium azide
CD300A
HPB acute lymphoid leukemia cell line (NM_007261.2).
Primary
This antibody (clone MEM-260) reacts with CD300a, a 60 kDa leukocyte transmembrane glycoprotein expressed on human granulocytes, monocytes, neutrophils, NK cells, mast cells and dendritic cells, 25% of circulating T cells and 15% of circulating B cells.
Store at 4°C in the dark.
Western Blot, Flow Cytometry
MEM-260
Western Blot, Flow Cytometry
CD300a antigenCMRF35H9, CD300a molecule, CLM-8, CMRF-35H, CMRF35-H, CMRF35H leukocyte immunoglobulin-like receptor, CMRF35-H9, CMRF-35-H9CD300 antigen-like family member A, CMRF35HIRp60, CMRF35-like molecule 8, IgSF12, IGSF12NK inhibitory receptor, Immunoglobulin superfamily member 12, Inhibitory receptor protein 60, IRC1, IRC1/IRC2, IRC2, Irp60, leukocyte membrane antigen
Mouse
0.1 mL
11314
Human
IgG1
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.