Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CD300a Monoclonal Antibody (172224)

Rat Monoclonal Antibody

Supplier:  Invitrogen™ MA523927

Catalog No. PIMA523927


Only null left
Add to Cart

Description

Description

In direct ELISAs, no cross-reactivity with recombinant human LMIR1, 2, 3, 4, 5, 6, recombinant mouse LMIR3, 4, or 5 is observed. Reconstitute at 0.5 mg/mL in sterile PBS.

The CMRF35 antigen (CMRF35A; MIM 606786), which was identified by reactivity with a monoclonal antibody, is present on monocytes, neutrophils, and some T and B lymphocytes. CMRF35H is recognized by the same antibody and is distinct from CMRF35 (Green et al., 1998 [PubMed 9701027]).
TRUSTED_SUSTAINABILITY
Specifications

Specifications

CD300a
Monoclonal
0.5 mg/mL
PBS with 5% trehalose and No Preservative
Q6SJQ0
CD300A
Mouse myeloma cell line NS0-derived recombinant mouse CD300a/LMIR1 Leu28-Arg183.
100 μg
Primary
Mouse
Antibody
IgG2a
Flow Cytometry
172224
Unconjugated
CD300A
B230315M08Rik; CD300 antigen-like family member A; Cd300a; CD300a antigen; CD300a molecule; Clm8; CLM-8; CMRF35H; CMRF-35H; CMRF35-H; CMRF35H leukocyte immunoglobulin-like receptor; CMRF35H9; CMRF-35-H9; CMRF35-H9; CMRF35HIRp60; CMRF35-like molecule 8; HSPC083; IGSF12; Immunoglobulin superfamily member 12; inhibitory receptor protein 60; IRC1; IRC1/IRC2; IRC2; IRp60; leukocyte membrane antigen; leukocyte mono-Ig-like receptor 1; Lmir1; MAIR-1; MAIR-I; MAIR-Ia; Mast cell-derived paired immunoglobulin-like receptor 1; Mcipr1; mcpir1; MMAC8; myeloid-associated immunoglobulin-like receptor 1; NK inhibitory receptor; NKRL; Pigr4; polymeric immunoglobulin receptor 4
Rat
Protein A/G
RUO
217303
-20°C, Avoid Freeze/Thaw Cycles
Lyophilized
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.