Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CD300a Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5103096
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5103096 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5103096 Supplier Invitrogen™ Supplier No. PA5103096
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total CD300A.

The CMRF35 antigen (CMRF35A; MIM 606786), which was identified by reactivity with a monoclonal antibody, is present on monocytes, neutrophils, and some T and B lymphocytes. CMRF35H is recognized by the same antibody and is distinct from CMRF35 (Green et al., 1998 [PubMed 9701027]).
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD300a
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene CD300A
Gene Accession No. Q9UGN4
Gene Alias B230315M08Rik; CD300 antigen-like family member A; Cd300a; CD300a antigen; CD300a molecule; Clm8; CLM-8; CMRF35H; CMRF-35H; CMRF35-H; CMRF35H leukocyte immunoglobulin-like receptor; CMRF35H9; CMRF-35-H9; CMRF35-H9; CMRF35HIRp60; CMRF35-like molecule 8; HSPC083; IGSF12; Immunoglobulin superfamily member 12; inhibitory receptor protein 60; IRC1; IRC1/IRC2; IRC2; IRp60; leukocyte membrane antigen; leukocyte mono-Ig-like receptor 1; Lmir1; MAIR-1; MAIR-I; MAIR-Ia; Mast cell-derived paired immunoglobulin-like receptor 1; Mcipr1; mcpir1; MMAC8; myeloid-associated immunoglobulin-like receptor 1; NK inhibitory receptor; NKRL; Pigr4; polymeric immunoglobulin receptor 4
Gene Symbols CD300A
Host Species Rabbit
Immunogen A synthesized peptide derived from human CD300A(Accession Q9UGN4), corresponding to amino acid residues K221-V271.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11314
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.