Learn More
Invitrogen™ CD300b Monoclonal Antibody (339003)

Description
In direct ELISAs, 100% cross-reactivity with recombinant human (rh) LMIR5 is observed and no cross-reactivity with recombinant mouse (rm) LMIR3, rmLMIR4, rhLMIR1, rhLMIR2, or rhLMIR6 is observed. Reconstitute at 0.5 mg/mL in sterile PBS.
Specifications
Specifications
| Antigen | CD300b |
| Applications | Western Blot |
| Classification | Monoclonal |
| Clone | 339003 |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 5% trehalose and No Preservative |
| Gene | CD300LB |
| Gene Accession No. | Q3U497 |
| Gene Alias | CD300 antigen like family member B; CD300 antigen-like family member B; CD300 molecule like family member b; CD300 molecule-like family member b; CD300b; CD300LB; CLM7; CLM-7; CMRF35A2; CMRF35-A2; CMRF35-like molecule 7; CMRF-35-like molecule-7; Gm253; Immune receptor expressed on myeloid cells 3; IREM3; IREM-3; Leukocyte mono-Ig-like receptor 5; LMIR5; TREM5; TREM-5; triggering receptor expressed on myeloid cells 5; UNQ2530/PRO6029 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.