Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD300c Antibody (105), Alexa Fluor™ 350 , Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP289993AF350
Description
CD300c Monoclonal antibody specifically detects CD300c in Human samples. It is validated for ELISASpecifications
CD300c | |
Monoclonal | |
Alexa Fluor 350 | |
CD300c antigenCMRF35 antigen, CD300c molecule, CLM-6, CMRF-35, CMRF35 leukocyte immunoglobulin-like receptor, CMRF-35A, CMRF35A leukocyte immunoglobulin-like receptor, CMRF35ACMRF35A1, CMRF35CMRF35-A1, CMRF35-like molecule 6, IgSF16, IGSF16CD300 antigen-like family member C, Immunoglobulin superfamily member 16, LIR | |
This antibody was obtained from a rabbit immunized with purified, recombinant Human CD300c (NP_006669.1; Met 1-Arg 183). | |
0.1 mL | |
Immunology | |
10871 | |
Store at 4°C in the dark. | |
IgG |
ELISA | |
105 | |
50mM Sodium Borate | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction