Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD300c Mouse, anti-Human, R718, Clone: TX45, BD OptiBuild™

Catalog No. BDB752008 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB752008 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB752008 Supplier BD Biosciences Supplier No. 752008
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The TX45 monoclonal antibody recognizes human CD300c which is also known as Leukocyte immunoglobulin-like receptor (LIR), CMRF35-like molecule 6 (CLM-6), or Immunoglobulin superfamily member 16 (IGSF16). CD300c is a type I transmembrane glycoprotein that belongs to the CMRF family within the Ig superfamily. CD300c is primarily expressed on myeloid cells and has also been detected on some T cells, B cells, and NK cells. Amongst freshly-isolated leucocytes, the TX45 antibody only detects CD300c expressed on monocytes. CD300c is involved in the regulation of proinflammatory cytokine production by leucocytes.

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD300c
Applications Flow Cytometry
Classification Monoclonal
Clone TX45
Conjugate R718
Host Species Mouse
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Refrigerate Upon Arrival
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.