Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CD300c Polyclonal Antibody

Catalog No. PIPA5122118
Change view
Click to view available options
Quantity:
20 μL
200 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5122118 20 μL
PIPA5119014 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. PIPA5122118 Supplier Invitrogen™ Supplier No. PA5122118
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

CD300c Polyclonal Antibody for Western Blot, IHC (P)

The CMRF35 antigen, which was identified by reactivity with a monoclonal antibody, is present on monocytes, neutrophils, and some T and B lymphocytes (Jackson et al., 1992 [PubMed 1349532]).
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD300c
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4
Gene CD300C
Gene Accession No. Q08708
Gene Alias CD300 antigen-like family member C; CD300c; CD300c antigen; CD300c molecule; Clm6; CLM-6; CMRF35; CMRF-35; CMRF35 antigen; CMRF35 leukocyte immunoglobulin-like receptor; CMRF35A; CMRF-35A; CMRF35A leukocyte immunoglobulin-like receptor; CMRF35A1; CMRF35-A1; CMRF-35-like molecule 6; CMRF35-like molecule 6; Gcom1; IgSF16; immunoglobulin superfamily member 16; LIR; RGD1565161
Gene Symbols CD300C
Host Species Rabbit
Immunogen Recombinant protein CD300 Antigen Like Family Member C. The antigen corresponds to amino acid range 21-183 of the target protein.
Purification Method Antigen affinity chromatography, Protein A
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100514248, 10871, 501745
Target Species Human, Pig, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.