Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CD300c Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA587884
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA587884 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA587884 Supplier Invitrogen™ Supplier No. PA587884
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: GYFPLSHPMT VAGPVGGSLS VQCRYEKEHR TLNKFWCRPP QILRCDKIVE TKGSAGKRNG RVSIRDSPAN LSFTVTLENL TEEDAGTYWC GVDTPWLRDF HDPIVEVEVS VFPAGTTTAS SPQSSMGTSG PPTKLPVHTW PSVTRKDSPE PSPHPGSLFS NVR; Positive Samples: Jurkat, Raji, A-549, PC-12, THP-1, Mouse heart, Mouse kidney; Cellular Location: Cell membrane, Single-pass type I membrane protein.

The CMRF35 antigen, which was identified by reactivity with a monoclonal antibody, is present on monocytes, neutrophils, and some T and B lymphocytes (Jackson et al., 1992 [PubMed 1349532]).
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD300c
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1.64 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene CD300C
Gene Accession No. A2A7V7, Q08708
Gene Alias CD300 antigen-like family member C; CD300c; CD300c antigen; CD300c molecule; Clm6; CLM-6; CMRF35; CMRF-35; CMRF35 antigen; CMRF35 leukocyte immunoglobulin-like receptor; CMRF35A; CMRF-35A; CMRF35A leukocyte immunoglobulin-like receptor; CMRF35A1; CMRF35-A1; CMRF-35-like molecule 6; CMRF35-like molecule 6; Gcom1; IgSF16; immunoglobulin superfamily member 16; LIR; RGD1565161
Gene Symbols CD300C
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 21-183 of human CD300C (NP_0066691).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10871, 387565, 501745
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.