Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD300e/LMIR6 Antibody (UP-H2), Janelia Fluor™ 646, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP262230JF646
Description
CD300e/LMIR6 Monoclonal specifically detects CD300e/LMIR6 in Human samples. It is validated for Western Blot, Flow Cytometry.Specifications
CD300e/LMIR6 | |
Monoclonal | |
Janelia Fluor 646 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
CD300E | |
CD300e-HA-transfected cells | |
0.1 mL | |
Primary | |
The mouse monoclonal antibody UP-H2 recognizes an extracellular epitope on CD300e/ IREM-2, a 32 kDa glycoprotein expressed by mature monocytes and peripheral blood myeloid dendritic cells. | |
Store at 4°C in the dark. | |
IgG1 |
Western Blot, Flow Cytometry | |
UP-H2 | |
Western Blot, Flow Cytometry | |
CD300 antigen like family member E, CD300 antigen-like family member E, CD300e antigenPIgR-2, CD300e molecule, CD300LEPIgR2, CLM-2, CLM2CMRF35-A5, CMRF35A5, CMRF35-like molecule 2, Immune receptor expressed on myeloid cells 2, IREM2IREM-2, Poly-Ig receptor 2, Polymeric immunoglobulin receptor 2 | |
Mouse | |
Protein A purified | |
Immunology | |
342510 | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction