Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD300f/LMIR3/CD300LF Antibody (234903), DyLight 594, Novus Biologicals™

Rat Monoclonal Antibody
Supplier: Novus Biologicals FAB2774M
Description
CD300f/LMIR3/CD300LF Monoclonal antibody specifically detects CD300f/LMIR3/CD300LF in Human samples. It is validated for Flow Cytometry, Immunocytochemistry, CyTOFSpecifications
CD300f/LMIR3/CD300LF | |
Monoclonal | |
DyLight 594 | |
50mM Sodium Borate | |
Rat | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
146722 | |
Store at 4°C in the dark. | |
IgG2b |
Flow Cytometry, Immunocytochemistry, CyTOF | |
234903 | |
Flow Cytometry, Immunocytochemistry, CyTOF-ready | |
CMRF35-like molecule 1, CD300 molecule-like family member f, CD300f, CLM1, CLM-1, IgSF13, IREM1, IREM-1, NKIR | |
Y3 rat myeloid cell line transfected with human CD300f/LMIR3 | |
0.1 mL | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction