Learn More
Invitrogen™ CD300f Monoclonal Antibody (234903)

Description
In direct ELISAs, no cross-reactivity with recombinant human LMIR1, 2, 4, 5, 6, or recombinant mouse LMIR3 is observed. Reconstitute at 0.5 mg/mL in sterile PBS.
Specifications
Specifications
| Antigen | CD300f |
| Applications | Flow Cytometry, Immunocytochemistry |
| Classification | Monoclonal |
| Clone | 234903 |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 5% trehalose and No Preservative |
| Gene | CD300LF |
| Gene Accession No. | Q8TDQ1 |
| Gene Alias | CD300 antigen like family member F; CD300 antigen-like family member F; CD300 molecule like family member f; CD300 molecule-like family member f; CD300f; Cd300lf; CLIM1; CLM1; CLM-1; CMRF-35-like molecule 1; CMRF35-like molecule 1; DC-derived Ig-like receptor 2; Digr2; F730004D16Rik; IgSF13; Immune receptor expressed on myeloid cells 1; immunoglobin superfamily member 13; immunoglobulin superfamily member 13; inhibitory receptor IREM1; IREM1; IREM-1; Leukocyte mono-Ig-like receptor 3; Lmir3; MAIR-5; MAIR-V; myeloid-associated immunoglobulin-like receptor 5; NK inhibitory receptor; NKIR; Pigr3; UNQ3105/PRO10111 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.