Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CD300f Monoclonal Antibody (2F1E11)
GREENER_CHOICE

Catalog No. FERMA548810
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FERMA548810 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FERMA548810 Supplier Invitrogen™ Supplier No. MA548810
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

CD300f Monoclonal Antibody for Western Blot, Flow, ELISA

CLM1 is an inhibitory receptor of the Ig superfamily expressed on myeloid cells. It mediates negative regulatory signals by recruiting SHP1 (PTPN6; MIM 176883) or SHIP.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD300f
Applications ELISA, Flow Cytometry, Western Blot, Western Blot
Classification Monoclonal
Clone 2F1E11
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% sodium azide
Gene CD300LF
Gene Accession No. Q8TDQ1
Gene Alias CD300 antigen like family member F; CD300 antigen-like family member F; CD300 molecule like family member f; CD300 molecule-like family member f; CD300f; Cd300lf; CLIM1; CLM1; CLM-1; CMRF-35-like molecule 1; CMRF35-like molecule 1; DC-derived Ig-like receptor 2; Digr2; F730004D16Rik; IgSF13; Immune receptor expressed on myeloid cells 1; immunoglobin superfamily member 13; immunoglobulin superfamily member 13; inhibitory receptor IREM1; IREM1; IREM-1; Leukocyte mono-Ig-like receptor 3; Lmir3; MAIR-5; MAIR-V; myeloid-associated immunoglobulin-like receptor 5; NK inhibitory receptor; NKIR; Pigr3; UNQ3105/PRO10111
Gene Symbols CD300LF
Host Species Mouse
Immunogen Purified recombinant fragment of human CD300F (AA: Extra(20-156)) expressed in E. Coli.
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 146722
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.