Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ CD300f Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA553080
Description
Immunogen sequence: SIVTQITGPT TVNGLERGSL TVQCVYRSGW EAYLKWWCRG AIWRDCKILV KTSGSEQEVK RDRVSIKDNQ KNRTFTVTME DLMKTDADTY WCGIEKTGND LGVTVQVTID PASTTASTPA PTTPTSTTFT APITQEETSS S Highest antigen sequence identity to the following orthologs: Mouse - 52%, Rat - 53%.
CLM1 is an inhibitory receptor of the Ig superfamily expressed on myeloid cells. It mediates negative regulatory signals by recruiting SHP1 (PTPN6; MIM 176883) or SHIP.
Specifications
CD300f | |
Polyclonal | |
Unconjugated | |
CD300LF | |
CD300 antigen like family member F; CD300 antigen-like family member F; CD300 molecule like family member f; CD300 molecule-like family member f; CD300f; Cd300lf; CLIM1; CLM1; CLM-1; CMRF-35-like molecule 1; CMRF35-like molecule 1; DC-derived Ig-like receptor 2; Digr2; F730004D16Rik; IgSF13; Immune receptor expressed on myeloid cells 1; immunoglobin superfamily member 13; immunoglobulin superfamily member 13; inhibitory receptor IREM1; IREM1; IREM-1; Leukocyte mono-Ig-like receptor 3; Lmir3; MAIR-5; MAIR-V; myeloid-associated immunoglobulin-like receptor 5; NK inhibitory receptor; NKIR; Pigr3; UNQ3105/PRO10111 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
146722 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.6 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q8TDQ1 | |
CD300LF | |
Recombinant protein corresponding to Human CD300f. Recombinant protein control fragment (Product #RP-90561). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction