Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CD300g Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA547755
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA547755 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA547755 Supplier Invitrogen™ Supplier No. PA547755
Add to Cart
Add to Cart

Sheep Polyclonal Antibody

In direct ELISAs, less than 1% cross-reactivity with recombinant human CD300g/Nepmucin is observed. Reconstitute at 0.2 mg/mL in sterile PBS.

CD300g, also known as Immunoglobulin-like Transcripts 4 (ILT4) or CMRF35-like molecule 1 (CLM-1), is a gene located on chromosome 17q25.1, encoding a member of the CD300 family of molecules involved in modulating immune responses. CD300g is expressed on myeloid cells, particularly macrophages and dendritic cells, and plays a role in regulating cell activation and inflammatory processes. It functions as an inhibitory receptor, interacting with phosphatidylserine and other lipid ligands to modulate immune cell activity, influencing cytokine production and phagocytosis. The expression and signaling pathways of CD300g contribute to immune homeostasis and are of particular interest in contexts like autoimmune diseases, infection, and cancer, where immune regulation is disrupted. Research into CD300g aims to uncover potential therapeutic applications in modulating immune functions and offers insights into its role in balancing immune responses in health and disease.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD300g
Applications Western Blot
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 5% trehalose and No Preservative
Gene CD300LG
Gene Accession No. Q1ERP8
Gene Alias 2310016B05Rik; CD300 antigen like family member G; CD300 antigen-like family member G; CD300 molecule like family member g; Cd300 molecule-like family member G; CD300g; CD300g antigen; Cd300lg; Clm9; CLM-9; CLM9Trem4; CMRF35-like molecule 9; CMRF-35-like molecule-9; D11Ertd736e; Nepmucin; RGD1311074; TREM4; TREM-4; Triggering receptor expressed on myeloid cells 4; UNQ422/PRO846
Gene Symbols CD300LG
Host Species Sheep
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse CD300g/Nepmucin Leu19-Ile284.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 52685
Target Species Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Lyophilized
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.