Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CD300d Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA572446
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA572446 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA572446 Supplier Invitrogen™ Supplier No. PA572446
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

CD300d Polyclonal Antibody for Western Blot

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD300d
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.09% sodium azide; pH 7.4
Gene CD300LD
Gene Accession No. Q6UXZ3, Q8VCH2
Gene Alias 4732429D16Rik; CD300 antigen like family member D; CD300 antigen-like family member D; CD300 molecule like family member d; CD300 molecule-like family member d; CD300D; CD300LD; CLM-4; CLM5; CLM-5; CMRF35A4; CMRF35-A4; CMRF35-like molecule 4; CMRF35-like molecule 5; immune receptor CD300d; leukocyte mono-Ig-like receptor 4; Lmir4; MAIR-4; MAIR-IV; myeloid-associated immunoglobulin (Ig)-like receptor IV; myeloid-associated immunoglobulin-like receptor 4; novel immunoglobulin V-set protein (Clm5); UNQ9218/PRO28686
Gene Symbols CD300LD
Host Species Rabbit
Immunogen region 76-115aa.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100131439, 217305
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.