Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD300LG/Nepmucin Antibody (102), mFluor Violet 450 SE, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP290215MFV450
Description
CD300LG/Nepmucin Monoclonal antibody specifically detects CD300LG/Nepmucin in Human samples. It is validated for ELISASpecifications
CD300LG/Nepmucin | |
Monoclonal | |
mFluor Violet 450 SE | |
50mM Sodium Borate | |
Rabbit | |
Protein A purified | |
RUO | |
146894 | |
Store at 4C in the dark. | |
IgG |
ELISA | |
102 | |
ELISA | |
CD300 antigen like family member G, CD300 antigen-like family member G, CD300 molecule like family member g, CD300 molecule-like family member g, CD300g antigen, CLM-9, CLM9Trem4, CMRF35-like molecule 9, NEPMUCIN, TREM4, TREM-4, Triggering receptor expressed on myeloid cells 4 | |
Rabbit immunized with purified, recombinant Human CD300LG/Nepmucin (Accession#: AAH25395.1, Met1-Arg247) | |
0.1 mL | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction