Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD300LG/Nepmucin Antibody, Janelia Fluor™ 549, Novus Biologicals™

Rabbit Polyclonal Antibody
$373.00
Specifications
Antigen | CD300LG/Nepmucin |
---|---|
Dilution | ELISA |
Classification | Polyclonal |
Conjugate | Janelia Fluor 549 |
Form | Purified |
Description
CD300LG/Nepmucin Polyclonal antibody specifically detects CD300LG/Nepmucin in Rat samples. It is validated for ELISASpecifications
CD300LG/Nepmucin | |
Polyclonal | |
Purified | |
RUO | |
CD300 antigen like family member G, CD300 antigen-like family member G, CD300 molecule like family member g, CD300 molecule-like family member g, CD300g antigen, CLM-9, CLM9Trem4, CMRF35-like molecule 9, NEPMUCIN, TREM4, TREM-4, Triggering receptor expressed on myeloid cells 4 | |
Produced in rabbits immunized with purified, recombinant Rat CD300LG/Nepmucin (Accession#: XP_002724611.1; Met1-Arg160). | |
Primary |
ELISA | |
Janelia Fluor 549 | |
Rabbit | |
50mM Sodium Borate | |
146894 | |
IgG | |
Antigen and protein A Affinity-purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title