Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD300LG Rabbit anti-Human, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
Supplier: Proteintech Group Inc 154261AP150UL

Description
CD300g receptor which may mediate L-selectin-dependent lymphocyte rollings. Binds SELL in a calcium dependent manner. Binds lymphocyte.Specifications
CD300LG | |
Polyclonal | |
Unconjugated | |
CD300LG | |
CD300LG, CLM 9, CLM9, CMRF35 like molecule 9, NEPMUCIN, TREM 4, TREM4 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
146894 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Western Blot | |
0.31 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q6UXG3 | |
CD300LG | |
CD300LG Fusion Protein Ag8075 | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction