Learn More
Invitrogen™ CD301 Monoclonal Antibody (744812), Alexa Fluor™ 488

Description
CD301 Monoclonal Antibody for Flow
Specifications
Specifications
| Antigen | CD301 |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | 744812 |
| Conjugate | Alexa Fluor 488 |
| Formulation | Buffered saline solution with ≤0.5% BSA and 0.09% sodium azide |
| Gene | CLEC10A |
| Gene Accession No. | Q8IUN9 |
| Gene Alias | CD301; CD301a; Clec10a; CLECSF13; CLECSF14; C-type (calcium dependent, carbohydrate-recognition domain) lectin, superfamily member 13 (macrophage-derived); C-type (calcium dependent, carbohydrate-recognition domain) lectin, superfamily member 14 (macrophage-derived); C-type lectin domain family 10 member A; C-type lectin domain family 10, member A; C-type lectin superfamily member 14; HML; HML2; macrophage asialoglycoprotein-binding protein 1; macrophage galactose N-acetyl-galactosamine specific lectin 1; Macrophage galactose/N-acetylgalactosamine-specific lectin; Macrophage lectin 2; macrophage lectin 2 (calcium dependent); M-ASGP-BP-1; MGL; Mgl1; MMGL |
| Gene Symbols | CLEC10A |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.