Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ CD301 Polyclonal Antibody
Rabbit Polyclonal Antibody
$209.50 - $511.50
Specifications
Antigen | CD301 |
---|---|
Concentration | 0.5 mg/mL |
Content And Storage | -20°C |
Applications | Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
Classification | Polyclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
PIPA5119033
|
Invitrogen™
PA5119033 |
200 μL |
Each for $511.50
|
|
|||||
PIPA5122117
|
Invitrogen™
PA5122117 |
20 μL |
Each for $209.50
|
|
|||||
Description
CD301 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P)
This gene encodes a member of the C-type lectin/C-type lectin-like domain (CTL/CTLD) superfamily. Members of this family share a common protein fold and have diverse functions, such as cell adhesion, cell-cell signalling, glycoprotein turnover, and roles in inflammation and immune response. The encoded type 2 transmembrane protein may function as a cell surface antigen. Two transcript variants encoding distinct isoforms have been identified for this gene.Specifications
CD301 | |
-20°C | |
Polyclonal | |
Liquid | |
IgG | |
Mouse | |
CLEC10A | |
CD301; CD301a; Clec10a; CLECSF13; CLECSF14; C-type (calcium dependent, carbohydrate-recognition domain) lectin, superfamily member 13 (macrophage-derived); C-type (calcium dependent, carbohydrate-recognition domain) lectin, superfamily member 14 (macrophage-derived); C-type lectin domain family 10 member A; C-type lectin domain family 10, member A; C-type lectin superfamily member 14; HML; HML2; macrophage asialoglycoprotein-binding protein 1; macrophage galactose N-acetyl-galactosamine specific lectin 1; Macrophage galactose/N-acetylgalactosamine-specific lectin; Macrophage lectin 2; macrophage lectin 2 (calcium dependent); M-ASGP-BP-1; MGL; Mgl1; MMGL | |
CLEC10A | |
Primary | |
Antigen affinity chromatography |
0.5 mg/mL | |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
Unconjugated | |
Rabbit | |
RUO | |
PBS with 50% glycerol and 0.05% ProClin 300 | |
P49300 | |
17312 | |
Recombinant protein Cluster Of differentiation 301. The antigen corresponds to amino acid range 69-273 of the target protein. | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title