Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CD301 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA582781
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA582781 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA582781 Supplier Invitrogen™ Supplier No. PA582781
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: CPVNWVEHQD SCYWFSHSGM SWAEAEKYCQ LKNAHLVVIN SREEQNFVQK YLGSAYTWMG LSDPEGAWKW VDGTDYATGF QNWKPGQPDD WQGHGLGGGE DCAHFHPDGR WND Highest antigen sequence indentity to the following orthologs: Mouse - 71%, Rat - 71%.

This gene encodes a member of the C-type lectin/C-type lectin-like domain (CTL/CTLD) superfamily. Members of this family share a common protein fold and have diverse functions, such as cell adhesion, cell-cell signalling, glycoprotein turnover, and roles in inflammation and immune response. The encoded type 2 transmembrane protein may function as a cell surface antigen. Two transcript variants encoding distinct isoforms have been identified for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD301
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.20 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene CLEC10A
Gene Accession No. Q8IUN9
Gene Alias CD301; CD301a; Clec10a; CLECSF13; CLECSF14; C-type (calcium dependent, carbohydrate-recognition domain) lectin, superfamily member 13 (macrophage-derived); C-type (calcium dependent, carbohydrate-recognition domain) lectin, superfamily member 14 (macrophage-derived); C-type lectin domain family 10 member A; C-type lectin domain family 10, member A; C-type lectin superfamily member 14; HML; HML2; macrophage asialoglycoprotein-binding protein 1; macrophage galactose N-acetyl-galactosamine specific lectin 1; Macrophage galactose/N-acetylgalactosamine-specific lectin; Macrophage lectin 2; macrophage lectin 2 (calcium dependent); M-ASGP-BP-1; MGL; Mgl1; MMGL
Gene Symbols CLEC10A
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human CLEC10A. Recombinant protein control fragment (Product #RP-88573).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10462
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.