Learn More
Invitrogen™ CD301 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA588266
Description
Immunogen sequence: FQNSKFQRDL VTLRTDFSNF TSNTVAEIQA LTSQGSSLEE TIASLKAEVE GFKQERQAGV SELQEHTTQK AHLGHCPHCP SVCVPVHSEM LLRVQQLVQD LKKLTCQVAT LNNNASTEGT C; Positive Samples: U-87MG, K-562, Raji, Mouse testis; Cellular Location: Membrane, Single-pass type II membrane protein.
This gene encodes a member of the C-type lectin/C-type lectin-like domain (CTL/CTLD) superfamily. Members of this family share a common protein fold and have diverse functions, such as cell adhesion, cell-cell signalling, glycoprotein turnover, and roles in inflammation and immune response. The encoded type 2 transmembrane protein may function as a cell surface antigen. Two transcript variants encoding distinct isoforms have been identified for this gene.
Specifications
CD301 | |
Polyclonal | |
Unconjugated | |
CLEC10A | |
CD301; CD301a; Clec10a; CLECSF13; CLECSF14; C-type (calcium dependent, carbohydrate-recognition domain) lectin, superfamily member 13 (macrophage-derived); C-type (calcium dependent, carbohydrate-recognition domain) lectin, superfamily member 14 (macrophage-derived); C-type lectin domain family 10 member A; C-type lectin domain family 10, member A; C-type lectin superfamily member 14; HML; HML2; macrophage asialoglycoprotein-binding protein 1; macrophage galactose N-acetyl-galactosamine specific lectin 1; Macrophage galactose/N-acetylgalactosamine-specific lectin; Macrophage lectin 2; macrophage lectin 2 (calcium dependent); M-ASGP-BP-1; MGL; Mgl1; MMGL | |
Rabbit | |
Affinity Chromatography | |
RUO | |
10462, 17312 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Western Blot | |
0.95 mg/mL | |
PBS with 50% glycerol and 0.05% ProClin 300; pH 7.3 | |
P49300, Q8IUN9 | |
CLEC10A | |
Recombinant fusion protein containing a sequence corresponding to amino acids 60-180 of human CD301/CLEC10A (NP_8789101). | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.