Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD301a/b (MGL1/2) Rat anti-Mouse , PE, Clone: ER-MP23, BD Biosciences™
Rat Monoclonal Antibody
$184.10 - $437.00
Specifications
| Antigen | CD301a/b (MGL1/2) |
|---|---|
| Clone | ER-MP23 |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Conjugate | PE |
Description
CD301a/b (MGL1/2) Monoclonal antibody specifically detects CD301a/b (MGL1/2) in Mouse samples. It is validated for Flow CytometrySpecifications
| CD301a/b (MGL1/2) | |
| Flow Cytometry | |
| PE | |
| RUO | |
| Aqueous buffered solution containing ≤0.09% sodium azide. | |
| Mouse Macrophage Precursor Cell Hybrids | |
| Primary |
| ER-MP23 | |
| Monoclonal | |
| Rat | |
| Mouse | |
| CD301a/CD301b; MGL1/MGL2; mMGL1/mMGL2 | |
| IgG2a κ | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated with R-PE under optimum conditions, and unconjugated antibody and free PE were removed. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title