Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ CD301a/b Polyclonal Antibody

Goat Polyclonal Antibody
Supplier: Invitrogen™ PA547609
Description
Reconstitute at 0.2 mg/mL in sterile PBS. Endoxin level is <0.10 EU per 1 μg of the antibody by the LAL method.
Specifications
| CD301a/b | |
| Polyclonal | |
| Unconjugated | |
| Clec10a | |
| CD301a; CD301b; Clec10a; C-type lectin domain family 10 member A; C-type lectin domain family 10, member A; Macrophage asialoglycoprotein-binding protein 1; macrophage galactose N-acetyl-galactosamine specific lectin 1; macrophage galactose N-acetyl-galactosamine specific lectin 2; Macrophage galactose/N-acetylgalactosamine-specific lectin; macrophage galactose-type C-type lectin 2; M-ASGP-BP-1; Mgl; Mgl1; Mgl2; MMGL | |
| Goat | |
| Antigen affinity chromatography | |
| RUO | |
| 17312, 216864 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
| Flow Cytometry, Neutralization, Western Blot | |
| 0.2 mg/mL | |
| PBS with 5% trehalose and No Preservative | |
| P49300 | |
| Clec10a, Mgl2 | |
| Mouse myeloma cell line NS0-derived recombinant mouse MGL1 Gln57-Ser304. | |
| 100 μg | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction