Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD301a (MGL1) Rat anti-Mouse, PE, Clone: LOM-8.7, BD Pharmingen™

Catalog No. BDB568900 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
0.1 mg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB568900 0.1 mg
BDB568901 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. BDB568900 Supplier BD Biosciences Supplier No. 568900
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rat Monoclonal Antibody

CD301a (MGL1) Monoclonal antibody specifically detects CD301a (MGL1) in Mouse samples. It is validated for Flow cytometry (Routinely Tested)
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD301a (MGL1)
Applications Flow Cytometry
Classification Monoclonal
Clone LOM-8.7
Concentration 0.2 mg/mL
Conjugate PE
Formulation Aqueous buffered solution containing protein stabilizer and ≤0.09% sodium azide.
Gene Alias Clec10a; CD301a; M-ASGP-BP-1; Mgl; Mgl1
Host Species Rat
Immunogen Mouse MGL1 Protein
Purification Method Affinity Chromatography
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Mouse
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated to the dye under optimum conditions and unconjugated antibody and free dye were removed.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.