Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CD301b (MGL2) Monoclonal Antibody (11A10-B7), eBioscience™, Invitrogen™

Catalog No. 14301137
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
2 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
14-301-137 2 mg
14-301-182 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 14-301-137 Supplier Invitrogen™ Supplier No. 14301137
Add to Cart
Add to Cart

Rat Monoclonal Antibody

Description: The monoclonal antibody 11A10-B7 recognizes mouse CD301b. CD301b is also known as macrophage galactose-type C-type lectin 2 (MGL2). CD301b expression is restricted to dermal dendritic cells, connection tissue, and bone marrow derived dendritic cells. Macrophages also express CD301b at low levels. CD301b contains a single carbohydrate recognition domain that recognizes protein antigens that have a galactose and N-acetylgalactosamine (Gal/GalNAc). Gal/GalNAc protein antigens are efficiently internalized through CD301b and presented to antigen specific CD4+ cells via MHC Class II. CD301b+ dendritic cells are believed to play an important role in TH2 priming. Applications Reported: This 11A10-B7 antibody has been reported for use in immunohistochemical staining of frozen tissue sections, and microscopy. Applications Tested: This 11A10-B7 antibody has been tested by immunohistochemistry of frozen mouse tissue and can be used at less than or equal to 10 μg/mL. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Purity: Greater than 90%, as determined by SDS-PAGE. Aggregation: Less than 10%, as determined by HPLC. Filtration: 0.2 μm post-manufacturing filtered.

CD301b is also known as macrophage galactose-type C-type lectin 2 (MGL2). CD301b expression is restricted to dermal dendritic cells, connection tissue, and bone marrow derived dendritic cells. Macrophages also express CD301b at low levels. CD301b contains a single carbohydrate recognition domain that recognizes protein antigens that have a galactose and N-acetylgalactosamine (Gal/GalNAc). Gal/GalNAc protein antigens are efficiently internalized through CD301b and presented to antigen specific CD4+ cells via MHC Class II. CD301b+ dendritic cells are believed to play an important role in TH2 priming.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD301b (MGL2)
Applications Immunohistochemistry (Frozen), Immunocytochemistry
Classification Monoclonal
Clone 11A10-B7
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.2
Gene Mgl2
Gene Accession No. 0
Gene Alias CD301; CD301b; macrophage galactose N-acetyl-galactosamine specific lectin 2; macrophage galactose-type C-type lectin 2; Mgl2
Gene Symbols Mgl2
Host Species Rat
Purification Method Affinity chromatography
Quantity 2 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 216864
Target Species Mouse
Content And Storage 4°C
Product Type Antibody
Form Liquid
Isotype IgG2a λ
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.