Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD301b (Mgl2) Rat anti-Mouse, BV421, Clone: URA-1, BD Horizon™

Catalog No. BDB569535 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
0.1 mg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB569535 0.1 mg
BDB569536 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BDB569535 Supplier BD Biosciences Supplier No. 569535
Add to Cart
Add to Cart

Rat Monoclonal Antibody

CD301b (Mgl2) Monoclonal antibody specifically detects CD301b (Mgl2) in Mouse samples. It is validated for Flow cytometry
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD301b (Mgl2)
Applications Flow Cytometry
Classification Monoclonal
Clone URA-1
Concentration 0.2 mg/mL
Conjugate Brilliant Violet 421
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Gene Alias CD301b; Mgl2; macrophage galactose-type C-type lectin 2
Host Species Rat
Immunogen Recombinant Mouse MGL2 Protein
Purification Method Affinity Chromatography
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 216864
Target Species Mouse
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated to the dye under optimum conditions and unconjugated antibody and free dye were removed.
Form Liquid
Isotype IgG2a λ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.