Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD302 Mouse anti-Human, Mouse, Clone: 2B1E8, Proteintech
SDP

Catalog No. 501737058 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-173-7058 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-173-7058 Supplier Proteintech Group Inc Supplier No. 666401IG150UL
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

CD302 is a C-type lectin receptor involved in cell adhesion and migration, as well as endocytosis and phagocytosis.

Specifications

Antigen CD302
Applications Western Blot
Classification Monoclonal
Clone 2B1E8
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Gene CD302
Gene Accession No. Q8IX05, Q9DCG2
Gene Alias BIMLEC, C type lectin BIMLEC, CD302, CD302 antigen, CD302 molecule, CLEC13A, DCL 1, DCL1, KIAA0022
Gene Symbols Cd302
Host Species Mouse
Immunogen CD302 Fusion Protein Ag9873
Purification Method Protein G
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 66205, 9936
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.