Learn More
Invitrogen™ CD303a Monoclonal Antibody (201A), APC, eBioscience™
Mouse Monoclonal Antibody
$467.00
Specifications
Antigen | CD303a |
---|---|
Clone | 201A |
Concentration | 5 μL/Test |
Content And Storage | 4°C, store in dark, DO NOT FREEZE! |
Applications | Flow Cytometry |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
17-981-842
![]() |
Invitrogen™
17981842 |
100 Tests |
Each of 1 for $467.00
|
|
|||||
Description
Description: This 201A monoclonal antibody reacts with human CD303a, which is also known as CLEC4C or Blood Dendritic Cell Antigen 2 (BDCA-2). This type II transmembrane protein is expressed almost exclusively on plasmacytoid dendritic cells, which secrete type I interferon during antiviral and antitumor responses. Studies have demonstrated that CD303a binds asialo-galactosyl-oligosaccharides on invading pathogens, resulting in the activation of signaling pathways that lead to CD303a internalization and downregulation of interferon production. Applications Reported: This 201A antibody has been reported for use in flow cytometric analysis. Applications Tested: This 201A antibody has been pre-titrated and tested by flow cytometric analysis of normal human peripheral blood cells. This can be used at 5 μL (0.125 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. Excitation: 633-647 nm; Emission: 660 nm; Laser: Red Laser. Filtration: 0.2 μm post-manufacturing filtered.
This gene encodes a member of the C-type lectin/C-type lectin-like domain superfamily. Members of this family share a common protein fold and have diverse functions, such as cell adhesion, cell-cell signalling, glycoprotein turnover, and roles in inflammation and immune response. The encoded type 2 transmembrane protein may play a role in dendritic cell function. Two transcript variants encoding distinct isoforms have been identified for this gene.Specifications
CD303a | |
5 μL/Test | |
Flow Cytometry | |
APC | |
Mouse | |
RUO | |
PBS with BSA and 0.09% sodium azide; pH 7.2 | |
Q8WTT0 | |
170482 | |
Primary | |
Affinity chromatography |
201A | |
4°C, store in dark, DO NOT FREEZE! | |
Monoclonal | |
Liquid | |
IgG2a κ | |
Human | |
CLEC4C | |
BDCA2; BDCA-2; Blood dendritic cell antigen 2; blood dendritic cell antigen 2 protein; CD303; CD303 antigen; CLEC4C; CLECSF11; CLECSF7; C-type (calcium dependent, carbohydrate-recognition domain) lectin, superfamily member 11; C-type (calcium dependent, carbohydrate-recognition domain) lectin, superfamily member 7; C-type lectin domain family 4 member C; C-type lectin domain family 4, member C; C-type lectin domain family 4, member C, isoform 1; C-type lectin superfamily member 7; dendritic cell lectin b; dendritic lectin; DLEC; HECL; PRO34150; UNQ9361/PRO34150 | |
CLEC4C | |
Antibody |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.