Learn More
Invitrogen™ CD304 (Neuropilin-1) Monoclonal Antibody (3DS304M), Super Bright™ 600, eBioscience™

Description
The monoclonal antibody 3DS304M recognizes mouse CD304 (Neuropilin-1), a type 1 transmembrane glycoprotein present on the surface of multiple cell types, including: regulatory T cells (Treg), NKT cells, DC, certain types of stem cells, neurons, endothelial cells, and some neoplastic cells. Neuropilin-1, in complex with plexins, serves as a co-receptor for type 3 semaphorins in growing neurons. It is also involved in the process of angiogenesis being a part of a functional receptor for VEGF in endothelial cells. In mice, Neuropilin-1 is expressed on thymus-derived natural Treg but not peripherally induced Treg. In addition, transient expression on recently activated non-regulatory T cells has been observed. Neuropilin-1 has been used as a marker of recent thymic emigrants in the mouse iNKT cell population. It has been shown that Neuropilin-1 forms a complex with TGF beta receptors, activating the latent form of TGF beta (LAP-TGF beta 1) and augmenting canonical Smad2/3 signaling. Applications Reported: This 3DS304M antibody has been reported for use in flow cytometric analysis. Applications Tested: This 3DS304M antibody has been tested by flow cytometric analysis of mouse splenocytes. This may be used at less than or equal to 0.5 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommende...
Specifications
Specifications
| Antigen | CD304 (Neuropilin-1) |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | 3DS304M |
| Concentration | 0.2 mg/mL |
| Conjugate | Super Bright 600 |
| Formulation | PBS with BSA and 0.09% sodium azide; pH 7.2 |
| Gene | NRP1 |
| Gene Accession No. | P97333 |
| Gene Alias | A5 protein; BDCA4; C530029I03; CD304; Neuropilin; neuropilin 1; Neuropilin1; neuropilin-1; Neuropilin-1 precursor (A5 protein); NP1; NP-1; Npn1; NPN-1; NRP; NRP 1; Nrp1; sNRP 1; sNRP1; soluble NRP 1; soluble NRP1; transmembrane receptor; vascular endothelial cell growth factor 165 receptor; VEGF165R |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.