Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CD305 (LAIR1) Monoclonal Antibody (113), PE, eBioscience™, Invitrogen™
GREENER_CHOICE

Catalog No. 5010749
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
50 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-107-49 100 μg
50-107-48 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-107-49 Supplier Invitrogen™ Supplier No. 12305182
Add to Cart
Edge
Add to Cart

Armenian Hamster Monoclonal Antibody

Description: CD305 or Leukocyte-associated Ig-like receptor-1 (LAIR-1) is a 46 kD cell surface molecule that functions as an inhibitory collagen-receptor on the majority of cells of the immune system. The mouse LAIR-1 molecule is expressed primarily on monocytes/macrophages and also on T cells, NK cells and dendritic cells. Unlike human B cells , mouse B cells do not express LAIR-1. The cytoplasmic tail of mouse LAIR-1 can be phosphorylated leading to the recruitment of SH2-containing tyrosine phosphatase(SHP)-2 and C-terminal Src kinase. Applications Reported: This 113 antibody has been reported for use in flow cytometric analysis. Applications Tested: This 113 antibody has been tested by flow cytometric analysis of mouse splenocytes. This can be used at less than or equal to 0.25 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Excitation: 488-561 nm; Emission: 578 nm; Laser: Blue Laser, Green Laser, Yellow-Green Laser. Filtration: 0.2 μm post-manufacturing filtered.

CD305, also known as LAIR1, is a type I transmembrane glycoprotein with an extracellular Ig-like domain and two intracellular ITIMs. It is expressed on a wide range of immune cells, including T, NK, and B cells, monocytes, dendritic cells, eosinophils, basophils, and mast cells. In humans, CD305 is also present on CD34+ hematopoietic progenitor cells and thymocytes. Several isoforms exist due to alternative splicing, with expression varying between cell types. CD305 functions as an inhibitory receptor, playing a key role in regulating immune responses. It blocks cytokine-mediated signaling, cell proliferation, and apoptosis. In NK, B, and T cells, CD305 modulates cytolytic functions and cytokine production, down-regulating IL2 and IFNG in CD4+ T cells while inducing transforming growth factor beta. In B cells, it reduces IgG and IgE production and decreases IL8, IL10, and TNF secretion. Activation of CD305 through tyrosine phosphorylation recruits phosphatases PTPN6 and PTPN11, although it may also function independently of SH2-containing phosphatases. It inhibits differentiation of peripheral blood precursors into dendritic cells and has been shown to induce apoptosis in myeloid leukemia cell lines. Collagens are identified as ligands for CD305, underscoring its role in immune regulation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD305 (LAIR1)
Applications Flow Cytometry
Classification Monoclonal
Clone 113
Concentration 0.2 mg/mL
Conjugate PE
Formulation PBS with 0.09% sodium azide; pH 7.2
Gene LAIR1
Gene Accession No. Q8BG84
Gene Alias 5133400O11Rik; BB115266; CD305; D7Bwg0421e; immunoglobulin heavy chain variable region; Lair1; Lair-1; leukocyte associated immunoglobulin like receptor 1; leukocyte-associated Ig-like receptor 1; leukocyte-associated Ig-like receptor-1; leukocyte-associated immunoglobulin-like receptor 1; rLAIR1
Gene Symbols LAIR1
Host Species Armenian Hamster
Purification Method Affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 52855
Target Species Mouse
Content And Storage 4°C, store in dark, DO NOT FREEZE!
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.