Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD307a (FCRL1) Mouse, anti-Human, R718, Clone: E3, BD OptiBuild™

Catalog No. BDB752120 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB752120 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB752120 Supplier BD Biosciences Supplier No. 752120
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The E3 monoclonal antibody specifically recognizes CD307a which is also known as Fc receptor-like protein 1 (FCRL1), Fc receptor homolog 1 (FCRH1), or Immune receptor translocation-associated protein 5 (IRTA5). CD307a (FCRL1) is a type I transmembrane glycoprotein that belongs to the IRTA gene family within the Ig gene superfamily. It has three extracellular Ig-like domains, a glutamic acid residue in its transmembrane region, and two consensus immunoreceptor tyrosine-based activation motifs (ITAMs) in its cytoplasmic domain. CD307a (FCRL1) bears sequence homology to classical Fc receptors. CD307a (FCRL1) is first expressed on pre-B cells and increases with maturation being expressed on naïve and memory B cells. It is downregulated by activated germinal center (GC) B cells. It may also be expressed by B cell-derived leukemia and lymphoma cells. CD307a may play key roles in B cell activation, differentiation and effector functions.

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD307a (FCRL1)
Applications Flow Cytometry
Classification Monoclonal
Clone E3
Conjugate R718
Host Species Mouse
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Refrigerate Upon Arrival
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.