Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD307c Mouse anti-Human, Brilliant Violet 650, Clone: H5, BD Optibuild™

Catalog No. BDB744238 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB744238 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB744238 Supplier BD Biosciences Supplier No. 744238
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The H5 monoclonal antibody specifically binds to CD307c which is also known as Fc receptor-like 3 (FCRL3), Immune receptor translocation-associated protein 3 (IRTA3), IFGP family protein 3 (IFGP3), and SH2 domain-containing phosphatase anchor protein 2 (SPAP2). The FCRL3 /gene is present in humans but not in mice. CD307c is a type I transmembrane glycoprotein that belongs to the FCRL family within the Ig gene superfamily. CD307c shares sequence homology with classical the Fc receptors. CD307c is expressed on subsets of B cells, plasma cells, NK cells, CD8+ T cells, and CD4+ natural T regulatory cells. CD307c isoforms contain multiple extracellular Ig domains and immunoreceptor-tyrosine activation (ITAM) and inhibitory (ITIM) motifs in their intracellular domains. CD307c may be involved in the regulation of immune responses. Genetic variation in FCRL3 /has been associated with susceptibility to certain autoimmune diseases.

The antibody was conjugated to BD Horizon™ BV650 which is part of the BD Horizon Brilliant™ Violet family of dyes. This dye is a tandem fluorochrome of BD Horizon BV421 with an Ex Max of 405nm and an acceptor dye with an Em Max at 650nm. BD Horizon BV650 can be excited by the violet laser and detected in a filter used to detect APC-like dyes (eg, 660/20nm filter). Due to the excitation and emission characteristics of the acceptor dye, there will be spillover into the APC and Alexa Fluor™ 700 detectors. However, the spillover can be corrected through compensation as with any other dye combination. BV650 is a tandem fluorochrome of BD Horizon BV421 and an acceptor dye with an emission maximum at 650 nm. Due to the excitation and emission characteristics of the acceptor dye, there will be spillover into the APC and Alexa Fluor™ 700 detectors. BV650 will have moderate spillover into the BD Horizon™ BV711 detector.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD307c
Classification Monoclonal
Clone H5
Concentration 0.2mg/mL
Conjugate Brilliant Violet 650
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Gene Alias FCRL3, Fc receptor-like 3, FcRH3, IRTA3, IFGP3, hIFGP3, SPAP2
Host Species Mouse
Immunogen Recombinant Human FCRL3
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG2b κ
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.