Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CD307e (FcRL5) Monoclonal Antibody (2A10D6)
GREENER_CHOICE

Catalog No. FERMA548789
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FERMA548789 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FERMA548789 Supplier Invitrogen™ Supplier No. MA548789
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

CD307e (FcRL5) Monoclonal Antibody for Western Blot, Flow, ELISA

Fc receptor homologs (FcRH5), also known as IRTA2 proteins, are transmembrane proteins with sequence homology to classical Fc receptor families. FcRH5 function as activating or inhibitory receptors on discrete B cell populations and are involved in B-cell development and differentiation in peripheral lymphoid organs. FcRH5 expression is deregulated in multiple myeloma and Burkitt lymphoma cell lines with 1q21 abnormalities. FcRH5 may play a critical role for the pathogenesis of B-cell lymphoma.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD307e (FcRL5)
Applications ELISA, Flow Cytometry, Western Blot, Western Blot
Classification Monoclonal
Clone 2A10D6
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% sodium azide
Gene FCRL5
Gene Accession No. Q96RD9
Gene Alias BXMAS1; BXMAS1-like protein 2; CD307; CD307e; fc receptor homolog 3; Fc receptor homolog 5; Fc receptor like 5; Fc receptor-like 3; Fc receptor-like 5; Fc receptor-like protein 3; Fc receptor-like protein 5; Fcrh3; FCRH5; Fcrl3; Fcrl5; FcR-like protein 5; Immune receptor translocation-associated protein 2; immunoglobulin superfamily receptor translocation associated 2 (IRTA2); IRTA2; mBXMH2; moFcRH3; PRO820; rCG62831-like; UNQ503/PRO820
Gene Symbols FCRL5
Host Species Mouse
Immunogen Purified recombinant fragment of human CD307E (AA: extra 16-158) expressed in E. Coli.
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 83416
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.