Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ CD307e (FcRL5) Monoclonal Antibody (307307)
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA524020
Description
In direct ELISAs, 100% cross-reactivity with recombinant human (rh) FCRL3 is observed and no cross-reactivity with rhFCRL1, 2, or 4 is observed. Reconstitute at 0.5 mg/mL in sterile PBS.
Fc receptor homologs (FcRH5), also known as IRTA2 proteins, are transmembrane proteins with sequence homology to classical Fc receptor families. FcRH5 function as activating or inhibitory receptors on discrete B cell populations and are involved in B-cell development and differentiation in peripheral lymphoid organs. FcRH5 expression is deregulated in multiple myeloma and Burkitt lymphoma cell lines with 1q21 abnormalities. FcRH5 may play a critical role for the pathogenesis of B-cell lymphoma.
Specifications
CD307e (FcRL5) | |
Monoclonal | |
0.5 mg/mL | |
PBS with 5% trehalose and No Preservative | |
Q96RD9 | |
FCRL5 | |
Mouse myeloma cell line NS0-derived recombinant human FCRL5/FCRL3 Gln16-Arg844. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG2b |
Flow Cytometry | |
307307 | |
Unconjugated | |
FCRL5 | |
BXMAS1; BXMAS1-like protein 2; CD307; CD307e; fc receptor homolog 3; Fc receptor homolog 5; Fc receptor like 5; Fc receptor-like 3; Fc receptor-like 5; Fc receptor-like protein 3; Fc receptor-like protein 5; Fcrh3; FCRH5; Fcrl3; Fcrl5; FcR-like protein 5; Immune receptor translocation-associated protein 2; immunoglobulin superfamily receptor translocation associated 2 (IRTA2); IRTA2; mBXMH2; moFcRH3; PRO820; rCG62831-like; UNQ503/PRO820 | |
Mouse | |
Protein A/G | |
RUO | |
83416 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction