Learn More
Invitrogen™ CD31 Monoclonal Antibody (390), PE-Cyanine7

Description
CD31 Monoclonal Antibody for Flow
Specifications
Specifications
| Antigen | CD31 |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | 390 |
| Concentration | 0.1 mg/mL |
| Conjugate | PE-Cyanine7 |
| Formulation | PBS with proprietary stabilizer and <0.1% sodium azide; pH 7.1 |
| Gene | Pecam1 |
| Gene Accession No. | Q08481 |
| Gene Alias | C85791; CD31; CD31 antigen; CD31, PECAM-1; CD31/EndoCAM; endoCAM; GPIIA'; I79_008304; PECA1; Pecam; PECAM1; Pecam-1; platelet and endothelial cell adhesion molecule 1; platelet endothelial cell adhesion molecule; platelet endothelial cell adhesion molecule-1; platelet/endothelial cell adhesion molecule (CD31 antigen); platelet/endothelial cell adhesion molecule 1; type I transmembrane endothelial adhesion molecule |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.