Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD31 Mouse anti-Human, Brilliant Violet 605, Clone: MBC 78.2 (also known as PECAM-1.2; 1.2), BD Optibuild™

Catalog No. BDB744076 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB744076 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB744076 Supplier BD Biosciences Supplier No. 744076
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The MBC 78.2 monoclonal antibody recognizes CD31 which is also known as, Platelet endothelial cell adhesion molecule (PECAM-1), platelet GPIIa, or EndoCAM. CD31 is a ñ130 kDa type I transmembrane glycoprotein that belongs to the Ig gene superfamily. CD31 is comprised of an extracellular region with six IgC-like domains, a transmembrane region, and a cytoplasmic domain that contains two immunoreceptor tyrosine-based inhibitory motifs (ITIMs). The MBC 78.2 antibody specifically binds to an epitope located on membrane-proximal, extracellular Ig-like domain 6 of CD31. This epitope remains expressed by activated T cells after enzymatic cleavage and shedding of a soluble extracellular CD31 fragment comprised of Ig-like domains 1 to 5 from cells. In contrast to the MBC 78.2 antibody, the WM59 monoclonal antibody reportedly binds to the extracellular Ig-like domain 2 of CD31. WM59 can thus bind to cells that express intact CD31 but not to cells that express a truncated form CD31 that lacks at least the membrane distal Ig-like domains 1 and 2 of CD31. CD31 has wide tissue distribution and is expressed on platelets, monocytes, granulocytes, some T cell subsets, and at high levels on endothelial cells. This cell adhesion molecule has been implicated in a number of cellular phenomena, including vascular wound healing. angiogenesis, transendothelial migration of leucocytes, and the regulation of T cell responses.

This antibody is conjugated to BD Horizon™ BV605 which is part of the BD Horizon Brilliant™ Violet family of dyes. With an Ex Max of 407nm and Em Max of 602nm, BD Horizon BV605 can be excited by a violet laser and detected with a standard 610/20nm filter set. BD Horizon BV605 is a tandem fluorochrome of BD Horizon BV421 and an acceptor dye with an Em max at 605nm. Due to the excitation of the acceptor dye by the green (532 nm) and yellow-green (561 nm) lasers, there will be significant spillover into the PE and BD Horizon PE-CF594 detectors off the green or yellow-green lasers. BD Horizon BV605 conjugates are very bright, often exhibiting brightness equivalent to PE conjugates and can be used as a third color off of the violet laser. BV605 is a tandem fluorochrome that combines BD Horizon BV421 and Cy™3.5. These conjugates are very bright, exhibiting similar brightness to equivalent PE conjugates. Due to the excitation of Cy3.5 by the green (532nm) and yellow-green (561nm) lasers, there will be significant spillover of the BD Horizon BV605 signal into the PE and BD Horizon PE-CF594 detectors off the green or yellow-green lasers.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD31
Classification Monoclonal
Clone MBC 78.2 (also known as PECAM-1.2; 1.2)
Concentration 0.2mg/mL
Conjugate Brilliant Violet 605
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Gene Alias PECAM-1, PECAM1, EndoCAM, GPIIA'
Host Species Mouse
Immunogen Human CD31
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG1 κ
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.