Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD31 Mouse, anti-Human, R718, Clone: M89D3, BD OptiBuild™

Catalog No. BDB752186 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB752186 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB752186 Supplier BD Biosciences Supplier No. 752186
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The M89D3 monoclonal antibody specifically recognizes CD31, which is also known as platelet endothelial cell adhesion molecule -1 (PECAM-1), a single chain, type I transmembrane protein of approximately 130-140 kDa. CD31 is a member of the immunoglobulin supergene family. Its extracellular region is composed of 574 amino acids and contains six C2-type Ig domains, and the intracellular region is composed of 118 residues. CD31 (PECAM-1) is expressed on endothelial cells, platelets, monocytes, neutrophils and NK cells. It has also been observed on subsets of T cells, but not on circulating B cells. It has been implicated in a number of cellular phenomena, including vascular wound healing, angiogenesis, and transendothelial migration of leukocytes and platelet aggregation in inflammatory responses.

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD31
Applications Flow Cytometry
Classification Monoclonal
Clone M89D3
Conjugate R718
Host Species Mouse
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Refrigerate Upon Arrival
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.