Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD31 Mouse Anti-Rat, Brilliant Violet 421, Clone: TLD-3A12, BD Optibuild™

Catalog No. BDB744357 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB744357 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB744357 Supplier BD Biosciences Supplier No. 744357
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The TLD-3A12 antibody specifically recognizes CD31, also known as PECAM-1 (platelet endothelial cell adhesion molecule). CD31 is a ñ130 kDa integral membrane glycoprotein, a member of the immunoglobulin superfamily, that mediates homophilic and heterophilic cell-cell adhesion. CD31 is expressed on endothelial cells, platelets, and subsets of leucocytes. In the human and mouse, multiple alternatively spliced isoforms have been identified, and this alternative splicing may be involved in the regulation of ligand specificity. CD31-mediated endothelial cell-cell interactions are involved in angiogenesis in the mouse and rat. In addition, PECAM-1 has been demonstrated to play an important role in extravasation of leucocytes in vivo, but in vivo treatment with TLD-3A12 mAb had no discernable effect upon the inflammatory infiltrate in experimental allergic encephalomyelitis. The TLD-3A12 mAb partially blocks the proliferative response of antigen-specific CD4+ T cells to antigen-presenting cells and relevant antigen. This CD31 antibody crossreacts with pig endothelial cells and platelets, as determined by immunohistochemical staining of acetone-fixed frozen sections and immunofluorescent staining with flow cytometric analysis, respectively. A suspension of Lewis rat microglial cells was used as the the immunogen.

The antibody was conjugated to BD Horizon™ BV421 which is part of the BD Horizon Brilliant™ Violet family of dyes. With an Ex Max of 407nm and Em Max at 421nm, BD Horizon BV421 can be excited by the violet laser and detected in the standard Pacific Blue™ filter set (eg, 450/50nm filter). BD Horizon BV421 conjugates are very bright, often exhibiting a 10 fold improvement in brightness compared to Pacific Blue conjugates. BV421 is a polymer-based dye excited by the violet laser and is one of the brightest fluorochromes offered by BD Biosciences. Conjugates are typically 10 times brighter than Pacific Blue™ conjugates and are often as bright as or brighter than PE conjugates. Due to nearly identical excitation and emission properties but different spillover characteristics, BD Horizon BV421, Pacific Blue™, and BD Horizon™ V450 cannot be used simultaneously.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD31
Classification Monoclonal
Clone TLD-3A12
Concentration 0.2mg/mL
Conjugate Brilliant Violet 421
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Gene Alias PECAM-1, Pecam1, Pecam
Host Species Mouse
Immunogen Lewis Rat Microglia
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Mouse
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG1 κ
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.