Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD31/PECAM-1.2 Mouse anti-Human, MilliporeSigma™

Catalog No. MABF203425 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABF2034-25 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABF2034-25 Supplier MilliporeSigma Supplier No. MABF203425UG
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-CD31/PECAM-1.2, clone MBC 78.2 is a mouse monoclonal that detects PECAM-1 and has been tested for use in ELISA, Flow Cytometry, Immunocytochemistry, Immunoprecipitation, Agonist or Inhibitor function, and Western Blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD31/PECAM-1.2
Applications ELISA, Flow Cytometry, Immunocytochemistry, Immunoprecipitation, Inhibition Assays, Western Blot
Classification Monoclonal
Concentration Please refer to lot specific datasheet.
Conjugate Unconjugated
Gene Symbols PECAM1
Host Species Mouse
Immunogen PECAM-1 purified from human platelets.
Purification Method Protein G Purified
Quantity 25 μg
Regulatory Status RUO
Research Discipline Inflammation and Immunology
Primary or Secondary Primary
Test Specificity Clone MBC 78.2 detects CD31/PECAM-1 in human cells. It targets an epitope within the IgG homology domain 6.
Target Species Human
Content And Storage Stable for 1 year at -20°C from date of receipt.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.