Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD31/PECAM-1 Antibody (693102), DyLight 405, Novus Biologicals™

Rat Monoclonal Antibody
Supplier: Novus Biologicals FAB3628E
Description
CD31/PECAM-1 Monoclonal antibody specifically detects CD31/PECAM-1 in Mouse samples. It is validated for Flow Cytometry, CyTOFSpecifications
CD31/PECAM-1 | |
Monoclonal | |
DyLight 405 | |
50mM Sodium Borate | |
Rat | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
Primary | |
Mouse | |
Purified |
Flow Cytometry, CyTOF | |
693102 | |
Flow Cytometry, CyTOF-ready | |
adhesion molecule, CD31, CD31 antigen, CD31/EndoCAM, EndoCAM, FLJ34100, FLJ58394, GPIIA', PECA1, PECAM-1, PECAM-1, CD31/EndoCAM, platelet endothelial cell adhesion molecule, platelet endothelial cell adhesion molecule-1, platelet/endothelial cell adhesion molecule | |
NS0 mouse myeloma cell line transfected with mouse CD31/PECAM-1, Glu18-Lys590, Accession # Q08481 | |
0.1 mL | |
Angiogenesis, Cancer, Cytoskeleton Markers, Embryonic Stem Cell Markers, Endothelial Cell Markers, Extracellular Matrix, Hematopoietic Stem Cell Markers, Immunology, Myeloid Cell Markers, Signal Transduction, Stem Cell Markers | |
5175 | |
Store at 4°C in the dark. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction