Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD31/PECAM-1 Antibody (C31.10), DyLight 550 , Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP311580R
Description
CD31/PECAM-1 Monoclonal antibody specifically detects CD31/PECAM-1 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
CD31/PECAM-1 | |
Monoclonal | |
DyLight 550 | |
adhesion molecule, CD31, CD31 antigen, CD31/EndoCAM, EndoCAM, FLJ34100, FLJ58394, GPIIA', PECA1, PECAM-1, PECAM-1, CD31/EndoCAM, platelet endothelial cell adhesion molecule, platelet endothelial cell adhesion molecule-1, platelet/endothelial cell adhesion molecule | |
Recombinant human CD31/PECAM-1 protein (Uniprot: P16284) | |
0.1 mL | |
Angiogenesis, Cancer, Cytoskeleton Markers, Embryonic Stem Cell Markers, Endothelial Cell Markers, Extracellular Matrix, Hematopoietic Stem Cell Markers, Immunology, Myeloid Cell Markers, Signal Transduction, Stem Cell Markers | |
5175 | |
Store at 4°C in the dark. | |
IgG1 κ |
Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
C31.10 | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction