Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD31/PECAM-1 Antibody (CO.3E-1D4) - BSA Free, Novus Biologicals™
SDP

Catalog No. p-7201047 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB017753 0.025 mg
NB100659001 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB017753 Supplier Novus Biologicals Supplier No. NB100659000.025MG
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody has been used in 6 publications

CD31/PECAM-1 Monoclonal specifically detects CD31/PECAM-1 in Bovine, Equine, Goat, Sheep samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.

Specifications

Antigen CD31/PECAM-1
Applications Immunohistochemistry (Frozen), Immunohistochemistry, Flow Cytometry, Immunoprecipitation
Classification Monoclonal
Clone CO.3E-1D4
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Flow Cytometry 1:10-1:25, Immunohistochemistry 1:50, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Frozen 1:50
Formulation Phosphate buffered saline with 0.09% Sodium Azide
Gene Alias adhesion molecule, CD31, CD31 antigen, CD31/EndoCAM, EndoCAM, FLJ34100, FLJ58394, GPIIA', PECA1, PECAM-1, PECAM-1, CD31/EndoCAM, platelet endothelial cell adhesion molecule, platelet/endothelial cell adhesion molecule
Gene Symbols PECAM1
Host Species Mouse
Immunogen Ovine leucocytes
Purification Method Protein G purified
Quantity 0.025 mg
Regulatory Status RUO
Research Discipline Cancer, Mesenchymal Stem Cell Markers, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 5175
Test Specificity NB100-65900 is specific for ovine CD31, also known as PECAM-1. Ovine CD31 is predominantly expressed by peripheral blood platelets and a small percentage of lymphocytes. CD31 is also highly expressed by ovine endothelial cells. Clone CO.3E1D4 is reported to inhibit homotypic leucocyte aggregation induced by anti CD43 antibodies (2). Removal of sodium azide is recommended prior to use in fuctional assays.
Target Species Goat, Sheep, Bovine
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2a
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.